Learn More
StatLab™ MasterTech Papanicolaou Stain Kit
SDP
Catalog No. STLKTPAPPT
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
STLKTPAPPT 1 pt.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTPAPPT Supplier StatLab™ Supplier No. KTPAPPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Papanicolaou Stain Kit stains atypical, inflammatory, and malignant cells in cytology smears in 15 minutes.
  • Both OG-6 and EA-50 serve as primary stains, producing producing pink, green, orange, yellow, or brown cytoplasm, pink to red nucleoli, cilia, squamous cells, and red blood cells
  • Hematoxylin acts as a secondary stain, producing blue chromatin and nuclei
  • 1 pt. Stain Kit yields between 320 to 500 slides
Specifications
Quantity 1 pt.
Product Type Stain Kit