Learn More
StatLab™ MasterTech PAS Stain Kit
SDP
Catalog No. STLKTPASPT
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTPASPT 1 pt.
STLKTPAS 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTPASPT Supplier StatLab™ Supplier No. KTPASPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
PAS Stain Kit stains for glycogen, mucin, fungi, and basement membranes in under 25 minutes.
  • Optimized Schiff's Solution serves as the primary stain, producing pink to red basement membranes, fungi, glycogen, and mucin against a green background, counterstained by light green
  • If Hematoxylin is used as a secondary stain, nuclei is rendered blue
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Quantity 1 pt.
Product Type Stain Kit