Learn More
StatLab MasterTech PAS Stain Kit
SDP
Catalog No. 21402143
Change view
Click to view available options
Product Type:
Periodic Acid Schiff
Catalog No. Product Type
21-402-143 Periodic Acid Schiff
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-143 Supplier StatLab Supplier No. KTPASLT
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
PAS stain kit for glycogen, mucin, fungi, and basement membranes in under 25 minutes
  • Optimized Schiff Solution serves as the primary stain, producing pink to red basement membranes, fungi, glycogen, and mucin against a green background, counterstained by light green
  • If Hematoxylin is used as a secondary stain, nuclei is rendered blue
  • Liter Stain Kit yields between 680-1,000 slides & slide yields assume maximum slides stained per run, maximum runs per use - Actual Slide Yields may vary
  • One or more component(s) in this kit require a storage temperature between 2°C and 10°C
  • Please review all component labels to ensure correct storage temperature
Specifications
Product Type Periodic Acid Schiff