Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Peanut Oil
SDP

Catalog No. 21033269
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-033-269 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-269 Supplier StatLab Supplier No. AHPOIPT
Add to Cart
Add to Cart

Used as a reagent to protect the acid-fastness of mycobacterial cells during the staining process

  • Used as a component in the Fite stain, a specialized staining technique used to identify leprosy bacteria (Mycobacterium leprae) within tissue samples
  • The oil is mixed with xylene to protect the bacteria's cell wall during deparaffinization, ensuring accurate staining results
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 8002-03-7
Color Yellow
Quantity 16 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.