Learn More
StatLab MasterTech Phloxine B
SDP
Catalog No. 21033457
Change view
Click to view available options
Quantity:
25 gm
Catalog No. Quantity
21-033-457 25 gm
1 options
Catalog No. 21-033-457 Supplier StatLab Supplier No. SPP0777
Used sed for its ability to stain specific cellular structures and demonstrate pathological changes
  • Solution is particularly effective in the Hematoxylin Phloxine Saffron (HPS) stain, where it provides a counterstain that differentiates cellular components and enhances diagnostic clarity
  • It has the ability to stain Paneth cell granules and demonstrate alcoholic hyaline underscores its utility in identifying specific cellular markers and pathological changes
  • It also has the versatility to act as a substitute for Eosin Y making it a preferred choice in various staining protocols
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 25 gm
Product Type Cytostain