Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Phosphate Buffer 6.8 pH
SDP

Catalog No. 22021376
Change view
Click to view available options
Quantity:
1 pt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-376 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-376 Supplier StatLab Supplier No. BUP1043
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Phosphate Buffer 6.8 pH, a MasterTech Special Stain Kit Component

  • Phosphate buffer is used with for the preservation of tissues and the preparation of staining solutions
  • It helps maintain the pH and osmolarity of the tissue samples, ensuring their viability and stability during processing
  • It is often used in conjunction with other fixatives to enhance the preservation of specific tissue types, such as the preservation of lipids with formal calcium formulation or the preservation of nuclear structures with divalent alkaline earth metals
  • Additionally, it can be supplemented with glucose or pyruvate to support cell viability and prevent the formation of formalin pigment
  • A MasterTech Special Stain Kit Component

Specifications

Grade Chemical Solution
Product Type Buffer Solution
Quantity 1 pt.
Solution Type Buffer
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.