Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Phosphate Buffered Saline
SDP

Catalog No. 21010264
Change view
Click to view available options
Grade:
Laboratory
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
21-010-264 Laboratory
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-264 Supplier StatLab Supplier No. BUP0257
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

A crucial reagent for washing cells and tissues.

  • Used to rinse cells and tissue samples to remove unwanted residues without causing harm to cellular structures
  • Can also be used for other histological purposes: a dilution buffer, transport medium, storage medium and for molecular cloning
  • A MasterTech Special Stain Kit Component

Specifications

For Use With (Equipment) Tissue Processors
Quantity 1 gal.
Solution Type Buffer
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.