Learn More
StatLab MasterTech Phosphotungstic/Phosphomolybdic Acid
SDP
Catalog No. 21010258
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-010-258 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-258 Supplier StatLab Supplier No. AHP0526
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Heteropoly acids for staining cell specimens.
  • They are components of the PTAH stain, which is used to stain connective tissue and other cellular structures
  • PTA is known for its ability to bind to fibrin, collagen, and fibres of connective tissues, selectively decolorizing them
  • A component for the following MasterTech Stain Kits - Massons Trichrome Stain Kit and Massons Trichrome 2000 Stain Kit
  • A MasterTech Special Stain Kit Component
    • They are components of the PTAH stain, which is used to stain connective tissue and other cellular structures
    • PTA is known for its ability to bind to fibrin, collagen, and fibres of connective tissues, selectively decolorizing them
    • 100 mL staining solution
    • A component for the following MasterTech Stain Kits - Massons Trichrome Stain Kit and Massons Trichrome 2000 Stain Kit
    • A MasterTech Special Stain Kit Component
Specifications
Quantity 100 mL
Product Type Phosphotungstic/Phosphomolybdic Acid