Learn More
StatLab MasterTech Picro-Sirius Red Pint Stain Kit
SDP
Catalog No. 22021397
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-021-397 1 pt.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-397 Supplier StatLab Supplier No. KTPSRPT
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Picro-Sirius Red Pint Stain Kit - Stains collagen and muscle in just over one hour
  • Oil Red O serves as the primary stain producing red fat with blue nuclei stained by the Hematoxylin
  • Pint Stain Kit yields between 320 to 500 slides and actual slide yields may vary
  • Pint Kit Consists of - Propylene Glycol 2 pints, Oil Red Stain O pint, and Modified Mayers Hematoxylin pint
  • All kit components are available for purchase individually and the kit is sealed with a tamper-evident label for quality assurance
  • All kit components should be stored in a dry environment between 15°C and 30°C
  • Please review all component labels to ensure correct storage temperature
  • A MasterTech Special Stain Kit
Specifications
Quantity 1 pt.
Product Type Picro-Sirius Red Stain