Learn More
StatLab MasterTech Picro-Sirius Red Stain

Description
- Used to detect and quantify collagen fibers in biological tissues
- It is particularly useful in the study of fibrosis, where excessive collagen deposition disrupts normal tissue function
- The stain provides precise visualization of collagen types I and III, making it essential for analyzing tissue architecture, fibrosis progression, and extracellular matrix (ECM) remodeling.
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Grade | Histological |
| Color | Red |
| Quantity | 1 pt. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.