Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Picroindigocarmine Solution
SDP

Catalog No. 21402139
Change view
Click to view available options
Quantity:
473 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-402-139 473 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-139 Supplier StatLab Supplier No. CUPICPT

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Solution for the selective staining of tissue structures

  • Solution is effective in highlighting cell nuclei and distinguishing certain structures, especially connective tissue and muscle
  • Solution consists of saturated picric acid and indigocarmine, with picric acid acting as a fixative and indigocarmine providing the characteristic coloration
  • This combination offers high sensitivity and specific binding to certain cellular components, making it a valuable tool in the preparation of cyto-histological samples for optical microscopy

Specifications

Physical Form Solution
Color Blue
CAS 88-89-1
Quantity 473 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.