Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Propylene Glycol
SDP

Catalog No. 22021369
Change view
Click to view available options
Quantity:
1 pt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-369 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-369 Supplier StatLab Supplier No. AHPGLPT
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Propylene Glycol, a MasterTech Special Stain Kit Component

  • Used as a solvent in the preparation of Oil Red O stain which specifically identifies and visualizes lipids (fat) within tissue sections particularly when examining frozen tissue samples
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 1 pt.
Grade Histological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.