Learn More
StatLab MasterTech Prostate Biopsy Fixative (NORT08)

Description
- Designed for tissue fixation in laboratory and histology workflows
- Helps preserve tissue morphology for subsequent processing and analysis
- Suitable for use in compatible laboratory tissue preparation protocols
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Histological |
| Product Type | Fixative |
| Solution Type | Fixative |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.