Learn More
StatLab MasterTech PTAH Stain Kit - Microwave
SDP
Catalog No. 23640010
Change view
Click to view available options
Quantity:
100 mL
473 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-010 473 mL
23-640-009 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-010 Supplier StatLab Supplier No. KTPTAPT
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Stains for collagen, fibrin, and striated muscle in under one hour
  • This kit includes a 55 minute microwave procedure and the traditional 15 hour procedure.
  • Unlike similar kits on the market, the MasterTech P.T.A.H. Stain Kit is less hazardous, using Zinc Chloride instead of mercuric based mordants, which also eliminates the need for Potassium Dichromate.
  • Ferric Ammonium Sulfate is also used instead of Acidic Potassium Permanganate, further reducing the stain kits hazard. P.T.A.H. serves as the primary stain, producing blue fibrin, muscle, and nuclei, with brown to red collagen.
  • A MasterTech Special Stain Kit.
Specifications
Quantity 473 mL
Product Type Special Stain Kit