Learn More
StatLab MasterTech Rhodanine Stock
SDP
Catalog No. 21010305
Change view
Click to view available options
Quantity:
25 mL
Catalog No. Quantity
21-010-305 25 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-305 Supplier StatLab Supplier No. KC132A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
For detecting copper deposits in tissue sections.
  • Rhodadine stock is primarily used in histology to detect copper deposits in tissue sections
  • The staining process involves the use of rhodadine, which binds with high affinity to copper-associated proteins, allowing for the visualization of copper deposits in various liver-related pathologies
  • This method is particularly useful in diagnosing conditions like Wilson's disease, where excess copper can lead to liver damage and other complications
  • A component for the MasterTech Microwave Copper Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 25 mL
Product Type Rhodadine Stock