Learn More
StatLab MasterTech Safranin O Cartilage Stain Kit
SDP
Catalog No. 21033407
Change view
Click to view available options
Volume (Metric):
16 oz.
Catalog No. Volume (Metric)
21-033-407 16 oz.
1 options
Catalog No. 21-033-407 Supplier StatLab Supplier No. KTSFOPT
Add to Cart
Add to Cart
Stains for cartilage in less than 15 minutes
  • This kit is commonly used for the staining of cartilage
  • 1% Safranin O serves as the primary stain, producing pink to red cartilage, mucin, and mast cell granules, with purple to black nuclei stained by the Weigert's Hematoxylin and cytoplasm counterstained green by Fast Green
  • Pint Stain Kit yields between 320-500 slides
  • Pint Kit Consists of: Weigert's Hematoxylin A pint, Weigert's Hematoxylin B pint, 0.2% Fast Green pint, 1% Safranin O pint, 1% Acetic Acid 2 pints
  • A MasterTech Special Stain Kit
Specifications
Volume (Metric) 16 oz.
Product Type Safranin O