Learn More
StatLab MasterTech Silver Nitrate Solution

Description
- A Staining agent to visualize delicate tissue structures, particularly nerve fibers, reticular fibers, and cellular components
- Solution acts by depositing metallic silver on tissues, making structures like neurofibrils, neurons, and fungal microorganisms visible under a microscope
- A Statlab MasterTech Special Stain Kit component
Specifications
Specifications
| Physical Form | Solution |
| CAS | 7761-88-8 |
| Color | Clear Colorless |
| Quantity | 500 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.