Learn More
MasterTech Sodium Borate Concentrate, StatLab

Description
- It is diluent in staining procedures, particularly for Hematoxylin and Eosin (H and E) and PASM/Jones stains
- It stabilizes pH to enhance staining, improves contrast in cellular components, and acts as a bactericidal preservative
- A StatLab MasterTech Special Stain Kit Component
Specifications
Specifications
| Physical Form | Solution |
| CAS | 1303-96-4 |
| Color | Colorless |
| Quantity | 30 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.