Learn More
StatLab MasterTech Sodium Metabisulfite Solution

Description
- Used as an antioxidant or reducing agent to stabilize dyes and prevent their oxidation during staining procedures thus improving the quality and durability of the stain by preserving its color and intensity
- A component for the MasterTech HelicoStat Rapid Helicobacter Stain Kit
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| For Use With (Equipment) | Staining Equipment |
| Grade | Chemical Solution |
| Product Type | Special Stain |
| Quantity | 1 pt. |
| Solution Type | Stain Kit |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.