Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, 0.2% Gold Chloride Solution
SDP

Catalog No. 23640137
Change view
Click to view available options
Color:
Clear Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-137 Clear Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-137 Supplier StatLab Supplier No. AHGCH0.2PT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

0.2% Gold Chloride Solution

  • 0.2% Gold Chloride Solution is a component for the following MasterTech Stain Kits: Jones Basement Membrane Stain Kit
  • Used for toning and stabilizing dye on tissue sections
  • Can be used as a toner by replacing some of the bound silver stain with gold can be used with various stains

Specifications

Physical Form Liquid
Color Clear Colorless
Quantity 1 p.
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.