Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, 10% Potassium Ferrocyanide
SDP

Catalog No. 23640146
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-146 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-146 Supplier StatLab Supplier No. AHP0850
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

10% Potassium Ferrocyanide

  • 10% Potassium Ferrocyanide can be used in some Iron Stain Procedures
  • Used for the detection of ferric iron
  • It combines with ferric ions in the tissue to form a bright blue pigment called Prussian blue
  • This method is used to differentiate collagen and muscle fibers in tissue sections and is especially helpful in studying pathologies of the heart, liver, and kidney

Specifications

Physical Form Aqueous Solution
Color Yellow
Quantity 1 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.