Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, 3% Acetic Acid
SDP

Catalog No. 23640130
Change view
Click to view available options
Color:
Clear Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-130 Clear Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-130 Supplier StatLab Supplier No. AHA0450
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

3% Acetic Acid

  • 3% Acetic Acid is a component for the following MasterTech Stain Kits: Alcian Blue pH1.0 Stain Kit, Alcian Blue pH2.5 Stain Kit, and the Russell-Movat Pentachrome Stain Kit
  • Used for staining both sulfated and carboxylated acid mucopolysaccharides and glycoproteins and removing excess blue and revealing different tissue elements.
  • Assists with demonstrating the presence of microorganisms in all types of tissues
  • For preserving nuclei but lysing red blood cells

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 1 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.