Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, 5% Oxalic Acid Solution
SDP

Catalog No. 23640176
Change view
Click to view available options
Color:
Colorless to Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-176 Colorless to Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-176 Supplier StatLab Supplier No. CUOAC5PT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

5% Oxalic Acid Solution

  • Used as a decontaminating agent due to its high acid content
  • Used as a liquefying agent to release mycobacterial cells from mucin in respiratory specimens
  • Used in staining for the identification of abnormalities in cell count and structure under the microscope

Specifications

Physical Form Aqueous Solution
Color Colorless to Yellow
Quantity 1 p.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.