Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab MasterTech Special Stain Kit, 5% Phosphotungstic Acid

Description
- A component for the following MasterTech Stain Kits: Heidenhains Aniline Blue-Azan Stain Kit and Russel-Movat Pentachrome Stain Kit
- Highlights muscle cross-striations and identifies fibrin
- Used for highlighting contraction bands in necrosis & showing gliosis in the central nervous system
- It is a polychrome stain that gives more than one color
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Clear Colorless |
| Quantity | 1 p. |
| Grade | Laboratory |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?