Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit Component, 10% Ferric Chloride
SDP

Catalog No. 23640136
Change view
Click to view available options
Quantity:
1 L
1 p.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-136 1 L
23-640-135 1 p.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-136 Supplier StatLab Supplier No. AHF0150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

10% Ferric Chloride

  • 10% Ferric Chloride is a component for the following MasterTech Stain Kits: Russell-Movat Pentachrome Stain Kit and Verhoeffs Elastic Stain Kit
  • Used to differentiate collagen and muscle fibers in tissue sections and study pathologies of the heart (infarction), liver (cirrhosis), and kidney (glomerular fibrosis)
  • Also used to detect deamination of phenylalanine in qualitative procedures and determine hippurate hydrolysis, phenylalanine deaminase, and tryptophan deaminase reactions in bacteria
  • Can be used to break the tissue-mordant dye complex and remove dye from the tissue

Specifications

Physical Form Aqueous Solution
Color Brown
Quantity 1 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.