Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit Component, 3% Methenamine Solution
SDP

Catalog No. 23640140
Change view
Click to view available options
Quantity:
1 L
1 p.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-140 1 p.
23-640-141 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-140 Supplier StatLab Supplier No. AHM0143
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

3% Methenamine Solution

  • A component for the MasterTech Jones Basement Membrane Reticulum Stain Kit
  • Used to visualize specific structures or microorganisms in tissue samples
  • Used to visualize basement membranes, primarily in kidney sections and identify dimorphic fungi Histoplasma spp. and yeast-like fungus Pneumocystis jiroveci, as well as differentiate fungal and bacterial cells

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 1 p.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.