Learn More
StatLab MasterTech Special Stain Kit Component, 5% Alcoholic Hematoxylin

Description
- 5% Alcoholic Hematoxylin is a component for the MasterTech Verhoeffs Elastic Stain Kit
- Used as an aggressive differentiating solution
- Sections are typically exposed to strong differentiating solutions for 2-10 seconds
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Brown |
| Quantity | 1 p. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.