Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit Component, 5% Silver Nitrate
SDP

Catalog No. 23640150
Change view
Click to view available options
Quantity:
1 p.
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-150 100 mL
23-640-151 1 p.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-150 Supplier StatLab Supplier No. AHS0426
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

5% Silver Nitrate

  • 5% Silver Nitrate is a component for the following MasterTech Stain Kits: Jones Basement Membrane Reticulum Stain Kit and Von Kossa Stain Kit
  • Used to visualize specific structures in tissue specimens due to its reducing properties and ability to interact with certain cellular components
  • Silver staining can also be used as a diagnostic tool for bacterial and fungal infections

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.