Learn More
StatLab MasterTech Special Stain Kit Component, 5% Silver Nitrate

Description
- 5% Silver Nitrate is a component for the following MasterTech Stain Kits: Jones Basement Membrane Reticulum Stain Kit and Von Kossa Stain Kit
- Used to visualize specific structures in tissue specimens due to its reducing properties and ability to interact with certain cellular components
- Silver staining can also be used as a diagnostic tool for bacterial and fungal infections
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Clear Colorless |
| Quantity | 1 p. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.