Learn More
StatLab MasterTech Special Stain Kit Component, Aniline Blue Stain
SDP
Catalog No. 23640132
Change view
Click to view available options
Quantity:
1 p.
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-132 1 p.
23-640-131 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-132 Supplier StatLab Supplier No. AHA0543
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Aniline Blue Stain
  • Used to stain collagen in the following MasterTech Stain Kits: Masson's Trichrome Stain Kit and Masson's Trichrome 2000 Stain Kit
  • Useful for identifying connective tissue and assessing fibrosis
Specifications
Quantity 1 p.
Product Type Stain Kit