Learn More
StatLab MasterTech Special Stain Kit Component, Biebrich Scarlet/Acid Fuchsin
SDP
Catalog No. 23640134
Change view
Click to view available options
Quantity:
1 p.
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-134 1 p.
23-640-133 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-134 Supplier StatLab Supplier No. AHB0443
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Biebrich Scarlet/Acid Fuchsin
  • Biebrich Scarlet/Acid Fuchsin is a component for the following MasterTech Stain Kits: Masson's Trichrome Stain Kit and Masson's Trichrome 2000 Stain Kit
  • An important histological stain used to differentiate various tissue components, particularly in connective tissue studies
  • Stains all acidophilic tissue components, including cytoplasm, muscle, and collagen
  • Used to differentiate between collagen and smooth muscle in tumors and to detect increased collagen in diseases such as cirrhosis
Specifications
Quantity 1 p.
Product Type Stain Kit