Learn More
StatLab MasterTech Special Stain Kit Component, Brown-Hopps 1% Crystal Violet

Description
- Brown-Hopps 1% Crystal Violet is a component for the MasterTech Brown-Hopps Stain Kit
- Bacteria appear on H and E as blue rods or cocci regardless of gram reaction
- Colonies appear as fuzzy blue clusters
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Blue |
| Quantity | 100 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.