Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit Component, Brown-Hopps 1% Crystal Violet
SDP

Catalog No. 23640194
Change view
Click to view available options
Quantity:
1 p.
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-194 1 p.
23-640-193 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-194 Supplier StatLab Supplier No. KC7912
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Brown-Hopps 1% Crystal Violet

  • Brown-Hopps 1% Crystal Violet is a component for the MasterTech Brown-Hopps Stain Kit
  • Bacteria appear on H and E as blue rods or cocci regardless of gram reaction
  • Colonies appear as fuzzy blue clusters

Specifications

Physical Form Aqueous Solution
Color Blue
Quantity 1 p.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.