Learn More
StatLab MasterTech Special Stain Kit Component, Phosphotungstic/Phosphomolybdic Acid

Description
- A component for the following MasterTech Stain Kits: Masson's Trichrome Stain Kit and Masson's Trichrome 2000 Stain Kit
- Staining specimens to enhance the contrast of biological samples.
- Used for assessing abnormalities in the liver, spleen, bone marrow, and kidneys and signaling the presence of tumors in the liver
- Diffuses out of collagen fibers to redden muscle cells
- Selectively stains muscle, collagen fibers, fibrin, and erythrocytes
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Yellow |
| Quantity | 1 p. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.