Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit Component, Phosphotungstic/Phosphomolybdic Acid
SDP

Catalog No. 23640144
Change view
Click to view available options
Quantity:
1 L
1 p.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-144 1 L
23-640-143 1 p.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-144 Supplier StatLab Supplier No. AHP0557
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Phosphotungstic/Phosphomolybdic Acid

  • A component for the following MasterTech Stain Kits: Masson's Trichrome Stain Kit and Masson's Trichrome 2000 Stain Kit
  • Staining specimens to enhance the contrast of biological samples.
  • Used for assessing abnormalities in the liver, spleen, bone marrow, and kidneys and signaling the presence of tumors in the liver
  • Diffuses out of collagen fibers to redden muscle cells
  • Selectively stains muscle, collagen fibers, fibrin, and erythrocytes

Specifications

Physical Form Aqueous Solution
Color Yellow
Quantity 1 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.