Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, Congo Red Stain
SDP

Catalog No. 23640260
Change view
Click to view available options
Quantity:
1 L
1 p.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-260 1 L
23-640-261 1 p.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-260 Supplier StatLab Supplier No. STCRELT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Congo Red Stain

  • A MasterTech Special Stain Kit Component
  • Congo red staining is a widely used histological technique for detecting amyloid deposits in tissue samples
  • Amyloids are abnormal protein aggregates associated with diseases such as Alzheimer's and systemic amyloidosis
  • The Congo Red dye is highly selective for amyloid deposits, which can lead to amyloidosis and also Alzheimer's

Specifications

Grade Laboratory
Color Red
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.