Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, Crocein Scarlet Acid Fuchsin
SDP

Catalog No. 23640189
Change view
Click to view available options
Color:
Purple/Violet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-189 Purple/Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-189 Supplier StatLab Supplier No. KC41B1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Crocein Scarlet Acid Fuchsin

  • Crocein Scarlet Acid Fuchsin is a component for the MasterTech Russell-Movat Pentachrome Stain Kit

Specifications

Physical Form Liquid
Color Purple/Violet
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.