Learn More
StatLab MasterTech Special Stain Kit, Eosin Y Stain, Aqueous
SDP
Catalog No. 23640262
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
23-640-262 1 L
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-262 Supplier StatLab Supplier No. STE0350
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Eosin Y Stain
  • A MasterTech H and E Stain
  • Non-hazardous and non-flammable alternative to traditional alcohol-based Eosin-Y formulations
  • MasterTech Aqueous Eosin-Y produces beautiful cytoplasmic staining while eliminating a source of hazard in the lab
Specifications
Quantity 1 L
Product Type Stain Kit