Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, Gold Chloride
SDP

Catalog No. 23640138
Change view
Click to view available options
Color:
Golden/Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-138 Golden/Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-138 Supplier StatLab Supplier No. AHGCH1100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Gold Chloride

  • Used as a toner in some histological silver stains converting metallic silver to metallic gold
  • For use in silver staining cuts to obtain better resolution, with a lighter background and a finer, more stable metal deposit
  • The staining process involves adding gold chloride, followed by removal of unreduced silver and excess gold chloride with sodium thiosulfate

Specifications

Physical Form Aqueous Solution
Color Golden/Green
Quantity 100 mL
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.