Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, HelicoStat
SDP

Catalog No. 23640195
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-195 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-195 Supplier StatLab Supplier No. KTHST

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

HelicoStat Special Stain Kit

  • Stains for Helicobacter pylori in 25 minutes
  • This kit is suitable in lieu of and comparable to Alcian Yellow/Toluidine Blue Helicobacter pylori Stains such as the Leung and Gibbon's Technique
  • The HelicoStat Stain Kit differs from these procedures by using true Alcian Yellow dye instead of an alternative, which provides tremendous contrast between the organism and mucin
  • Toluidine Blue serves as the primary stain, producing blue Helicobacter pylori and background against yellow mucins, counterstained by Alcian Yellow
  • 100 mL Stain Kit yields between 53 and 66 slides
  • 100 mL Kit Consists of: Periodic Acid Solution Pint, Alcian Yellow Solution 100ml, 3% Sodium Hydroxide 10 mL, Sodium Metabisulfite Solution 100 mL and Toluidine Blue Stock

Specifications

Physical Form Aqueous Solution
Color Blue
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.