Learn More
StatLab MasterTech Special Stain Kit, Oil Red O Stain
SDP
Catalog No. 23640263
Change view
Click to view available options
100 mL
Catalog No.
23-640-263 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-263 Supplier StatLab Supplier No. STORO100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Oil Red O Stain
  • A MasterTech Special Stain Kit Component
  • Oil Red O (ORO) is a fat-soluble dye commonly used in histology to detect and quantify lipid content in cells and tissues
  • It specifically stains neutral lipids, such as triglycerides and cholesteryl esters, a bright red color, which makes it a valuable tool for diagnosing conditions such as fatty liver disease
Specifications
Quantity 100 mL
Product Type Stain Kit