Learn More
StatLab MasterTech Special Stain Kit, One Step Trichrome Stain (Green and Red)
SDP
Catalog No. 23640266
Change view
Click to view available options
Quantity:
1 p.
Catalog No. Quantity
23-640-266 1 p.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-266 Supplier StatLab Supplier No. STOSTGPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
One Step Trichrome Stain (Green and Red)
  • A MasterTech Special Stain Kit Component
  • One Step Trichrome Stain (Green and Red) is used in histology lab applications to visualize tissue architecture
  • It provides distinct color contrasts, allowing researchers and pathologists to see collagen, muscle fibers, and nuclei in tissue samples
Specifications
Quantity 1 p.
Product Type Stain Kit