Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, Picric Acid Acetone (Traditional Formulation)
SDP

Catalog No. 23640267
Change view
Click to view available options
:
1 Pint
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-640-267 1 Pint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-267 Supplier StatLab Supplier No. STPAAPT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Picric Acid Acetone (Traditional Formulation)

  • A MasterTech Special Stain Kit Component
  • Picric Acid Acetone Traditional Formulation
  • Used to differentiate the safranin and counterstain the background

Specifications

Physical Form Liquid
Color Yellow
Quantity 1 p.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.