Learn More
StatLab MasterTech Special Stain Kit, PTAH Stain, Phosphotungstic Acid Hematoxylin Stain
SDP
Catalog No. 23640268
Change view
Click to view available options
Quantity:
1 p.
Catalog No. Quantity
23-640-268 1 p.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-268 Supplier StatLab Supplier No. STPTAPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
PTAH Stain - Phosphotungstic Acid Hematoxylin Stain
  • A MasterTech Special Stain Kit Component
  • Phosphotungstic Acid Hematoxylin (PTAH) is a histological stain used to demonstrate collagen, muscle striations, and central nervous system structures
  • It binds to fibrin, collagen, and fibres of connective tissues, and replaces the anions of dyes from these materials, selectively decoloring them
  • PTAH is used to show gliosis in the central nervous system, tumors of skeletal muscles, and fibrin deposits in lesions
Specifications
Quantity 1 p.
Product Type Stain Kit