Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, Sodium Chloride Crystals
SDP

Catalog No. 23640258
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-258 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-258 Supplier StatLab Supplier No. SPS0985
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Sodium Chloride Crystals

  • A MasterTech Special Stain Kit Component
  • Sodium Chloride Crystals are used in histology lab applications for tissue preservation, influencing staining outcomes, and aiding in the efficient extraction of high-quality DNA and RNA

Specifications

Physical Form Crystalline Powder
Color White
Quantity 500 g
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.