Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, Sodium Phosphate Monobasic Anhydrous
SDP

Catalog No. 23640259
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-259 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-259 Supplier StatLab Supplier No. SPS2085
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Sodium Phosphate Monobasic Anhydrous

  • A MasterTech Special Stain Kit Component
  • Used as a component of phosphate buffer saline (PBS buffer) used in the study of glucose biosensor
  • Used in buffers for molecular biology applications
  • Also used in cholesterol assay reaction buffer and miRNA microarray fabrication

Specifications

Physical Form Crystalline Powder
Color White
Quantity 500 g
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.