Learn More
StatLab MasterTech Special Stain Kit, Universal Iodine Solution
SDP
Catalog No. 23900135
Change view
Click to view available options
1 Pint
Catalog No.
23-900-135 1 Pint
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-900-135 Supplier StatLab Supplier No. STUIOPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Universal Iodine Solution
  • A MasterTech Special Stain Kit Component
  • Universal Iodine Solution works for all histology applications and is a component in the following MasterTech Stain Kits: MasterTech Brown and Brenn Modified Stain Kit, MasterTech Verhoeff's Elastic Stain Kit, MasterTech McDonald's Gram Stain Kit™, and MasterTech Russell-Movat Pentachrome Stain Kit
  • Universal iodine solution is used in histology for staining cell structures
  • It enhances contrast and makes cell structures more visible under the microscope
Specifications
Quantity 1 p.
Product Type Gram Iodine