Learn More
StatLab MasterTech Special Stain Kit, Van Gieson's Stain
SDP
Catalog No. 23640269
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
23-640-269 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-269 Supplier StatLab Supplier No. STVGI100
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Van Gieson's Stain
  • A MasterTech Special Stain Kit Component
  • Van Gieson's Stain is used for differentiating collagen fibers from muscle and cytoplasm in tissue samples
  • It is primarily used for identifying and assessing elastic fibers in tissues such as skin, lungs, and blood vessels, as well as diagnosing vascular diseases and examining connective tissue disorders
Specifications
Quantity 100 mL
Product Type Stain Kit