Learn More
StatLab MasterTech Stain Retrieval Solution
SDP
Catalog No. 21010371
Change view
Click to view available options
Form:
Solution
Catalog No. Form
21-010-371 Solution
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-371 Supplier StatLab Supplier No. STZ00388E
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Designed to help bring back damaged tissue stain sites so H and E stains, special stains, and immunostains look excellent again.
  • For those tissues that got damaged along the way - excessive heat, formalin crosslinking, and inadequate tissue processing may cause stain dysfunction
  • A MasterTech Special Stain Kit Component
Specifications
Product Type Retrieval Solution
Solution Type Stain Retrieval Solution
Form Solution