Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Steiner Stain Kit
SDP

Catalog No. 22046644
Change view
Click to view available options
Format:
Pint
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22-046-644 Pint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-046-644 Supplier StatLab Supplier No. KTSTEPT
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Stains for Donovan bodies, fungi, Helicobacter pylori, legionella pneumophila, and spirochetes in 45 minutes.

  • Kit includes a 45 minute microwave procedure and the traditional one hour procedure.
  • Differs from similar kits by eliminating radioactive Uranyl Nitrate, providing pre-measured Hydroquinone capsules, and modifying the original technique, which creates a safer, easier to use procedure that provides optimal staining results and reduced background staining.
  • Silver Nitrate, developed by the Hydroquinone based reducing solution serves as the primary stain, producing dark brown to black donovan bodies, fungi, Helicobacter pylori, legionella pneumophila, and spirochetes, against a yellow to brown background.
  • Kit yields between 50 to 192 slides.

Specifications

Assay 50 to 196 Slides
Format Pint
For Use With (Application) Donovan bodies, funig, Helicobacter Pylori, Legionella pneumophila, spirochetes
Product Type Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.